missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3-methylbenzoic Acid 98.0+%, TCI America™

Catalog No. H13195G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H13195G 5 g
H131925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H13195G Supplier TCI America Supplier No. H13195G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 499-76-3
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00270105
InChI Key LTFHNKUKQYVHDX-UHFFFAOYSA-N
Synonym 4-hydroxy-m-toluic acid, 3-methyl-4-hydroxybenzoic acid, 4-hydroxy-3-methyl-benzoic acid, 4,3-cresotic acid, 4-hydroxy-3-methylbenzoate, benzoic acid, 4-hydroxy-3-methyl, 4-hydroxy-m-toluate, pubchem20044, acmc-209kj4, 3-methyl-4-hydroxybenzoate
PubChem CID 68138
ChEBI CHEBI:85239
IUPAC Name 4-hydroxy-3-methylbenzoic acid
SMILES CC1=CC(=CC=C1O)C(O)=O

Specifications

Melting Point 175°C
Quantity 5 g
Formula Weight 152.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxy-3-methylbenzoic Acid