missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3-methyl-2-butanone, tech 85%

Catalog No. AAH2848303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2848303 1 g
AAH2848306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2848303 Supplier Thermo Scientific Chemicals Supplier No. H2848303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3393-64-4 | C5H10O2 | 102.133 g/mol

Protonated diacetone alcohol and protonated 4- hydroxy-3-methyl-2-butanone undergo vinylation and colli- sion-induced dehydration as the only primary reactions, the product of the former reaction consisting of up to 40% of the total product ion distribution.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Protonated diacetone alcohol and protonated 4- hydroxy-3-methyl-2-butanone undergo “vinylation” and colli- sion-induced dehydration as the only primary reactions, the product of the former reaction consisting of up to 40% of the total product ion distribution.

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 3393-64-4
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.133
MDL Number MFCD00004739
InChI Key VVSRECWZBBJOTG-UHFFFAOYSA-N
Synonym 4-hydroxy-3-methyl-2-butanone, 2-butanone, 4-hydroxy-3-methyl, 3-hydroxymethyl-2-butanone, 2-butanone, 4-hydroxy-3-methyl-, +, acmc-1cngd, 3-methyl-4-hydroxy-2-butanone, vvsrecwzbbjotg-uhfffaoysa, 2-butanone,4-hydroxy-3-methyl, 4-hydroxy-3-methyl-2-butanone, technical grade
PubChem CID 18829
IUPAC Name 4-hydroxy-3-methylbutan-2-one
SMILES CC(CO)C(=O)C

Specifications

Refractive Index 1.4325
Quantity 1 g
Formula Weight 102.13
Percent Purity 85%
Grade Technical
Chemical Name or Material 4-Hydroxy-3-methyl-2-butanone
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 222-238-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only