accessibility menu, dialog, popup

UserName

CAS RN 3393-64-4

IUPAC Name: 4-hydroxy-3-methylbutan-2-one
Synonyms: 4-hydroxy-3-methyl-2-butanone
Molecular Weight (g/mol): 102.13
Molecular Formula: C5H10O2
InChi Key: VVSRECWZBBJOTG-UHFFFAOYNA-N
SMILES: CC(CO)C(C)=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results