missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3-methoxyphenylacetic Acid 98.0+%, TCI America™

Catalog No. H03391G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H03391G 1 g
H03395G 5 g
H033925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H03391G Supplier TCI America Supplier No. H03391G

Chemical Identifiers

CAS 306-08-1
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.175
MDL Number MFCD00004350
InChI Key QRMZSPFSDQBLIX-UHFFFAOYSA-N
Synonym homovanillic acid, 4-hydroxy-3-methoxyphenylacetic acid, 2-4-hydroxy-3-methoxyphenyl acetic acid, vanillacetic acid, benzeneacetic acid, 4-hydroxy-3-methoxy, 3-methoxy-4-hydroxyphenylacetic acid, homovaniuic acid, 4-hydroxy-3-methoxyphenyl acetic acid, homovanillate, 4-hydroxy-3-methoxybenzeneacetic acid
PubChem CID 1738
ChEBI CHEBI:545959
IUPAC Name 2-(4-hydroxy-3-methoxyphenyl)acetic acid
SMILES COC1=C(C=CC(=C1)CC(=O)O)O

Specifications

Melting Point 143°C
Quantity 1 g
Formula Weight 182.18
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxy-3-methoxyphenylacetic Acid