missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Hydroxy-3-methoxybenzonitrile 98.0+%, TCI America™
$86.64 - $338.84
Chemical Identifiers
| CAS | 4421-08-3 |
|---|---|
| Molecular Formula | C8H7NO2 |
| Molecular Weight (g/mol) | 149.15 |
| MDL Number | MFCD00001820 |
| InChI Key | QJRWLNLUIAJTAD-UHFFFAOYSA-N |
| Synonym | vanillonitrile, m-anisonitrile, 4-hydroxy, 4-hydroxy-3-methoxy-benzonitrile, 3-methoxy-4-hydroxybenzonitrile, 4-hydroxy-3-methoxybenzenecarbonitrile, pubchem13112, 2-methoxy-4-cyanophenol, 4-cyano-2-methoxyphenol, 5-cyano-2-hydroxyanisole, 2-methoxy-4-cyano-phenol |
| PubChem CID | 78135 |
| IUPAC Name | 4-hydroxy-3-methoxybenzonitrile |
| SMILES | COC1=C(C=CC(=C1)C#N)O |
Chemical Identifiers
| 4421-08-3 | |
| 149.15 | |
| QJRWLNLUIAJTAD-UHFFFAOYSA-N | |
| 78135 | |
| COC1=C(C=CC(=C1)C#N)O |
| C8H7NO2 | |
| MFCD00001820 | |
| vanillonitrile, m-anisonitrile, 4-hydroxy, 4-hydroxy-3-methoxy-benzonitrile, 3-methoxy-4-hydroxybenzonitrile, 4-hydroxy-3-methoxybenzenecarbonitrile, pubchem13112, 2-methoxy-4-cyanophenol, 4-cyano-2-methoxyphenol, 5-cyano-2-hydroxyanisole, 2-methoxy-4-cyano-phenol | |
| 4-hydroxy-3-methoxybenzonitrile |
Specifications
| 4421-08-3 | |
| C8H7NO2 | |
| 5 g | |
| vanillonitrile, m-anisonitrile, 4-hydroxy, 4-hydroxy-3-methoxy-benzonitrile, 3-methoxy-4-hydroxybenzonitrile, 4-hydroxy-3-methoxybenzenecarbonitrile, pubchem13112, 2-methoxy-4-cyanophenol, 4-cyano-2-methoxyphenol, 5-cyano-2-hydroxyanisole, 2-methoxy-4-cyano-phenol | |
| COC1=C(C=CC(=C1)C#N)O | |
| 149.15 | |
| 149.15 | |
| Crystalline Powder |
| 87°C | |
| MFCD00001820 | |
| 3439 | |
| QJRWLNLUIAJTAD-UHFFFAOYSA-N | |
| 4-hydroxy-3-methoxybenzonitrile | |
| 78135 | |
| ≥98.0% (GC,T) | |
| 4-Hydroxy-3-methoxybenzonitrile |
Safety and Handling
TSCA : No