missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3-methoxybenzonitrile, 98%

Catalog No. AAA1805614
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1805614 25 g
AAA1805622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1805614 Supplier Thermo Scientific Chemicals Supplier No. A1805614

CAS: 4421-08-3 | C8H7NO2 | 149.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4421-08-3
MDL Number MFCD00001820

Specifications

Melting Point 85°C to 89°C
Molecular Formula C8H7NO2
Quantity 25 g
UN Number UN3439
Synonym vanillonitrile, m-anisonitrile, 4-hydroxy, 4-hydroxy-3-methoxy-benzonitrile, 3-methoxy-4-hydroxybenzonitrile, 4-hydroxy-3-methoxybenzenecarbonitrile, pubchem13112, 2-methoxy-4-cyanophenol, 4-cyano-2-methoxyphenol, 5-cyano-2-hydroxyanisole, 2-methoxy-4-cyano-phenol
InChI Key QJRWLNLUIAJTAD-UHFFFAOYSA-N
SMILES COC1=C(C=CC(=C1)C#N)O
IUPAC Name 4-hydroxy-3-methoxybenzonitrile
Molecular Weight (g/mol) 149.15
PubChem CID 78135
Formula Weight 149.15
Percent Purity 98%
Chemical Name or Material 4-Hydroxy-3-methoxybenzonitrile
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 224-589-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only