Learn More
4-Hydroxy-3-methoxybenzoic acid, 97+%
CAS: 121-34-6 | C8H8O4 | 168.15 g/mol
$79.56 - $79.56
Chemical Identifiers
| CAS | 121-34-6 |
|---|---|
| Molecular Formula | C8H8O4 |
| Molecular Weight (g/mol) | 168.15 |
| MDL Number | MFCD00002551 |
| InChI Key | WKOLLVMJNQIZCI-UHFFFAOYSA-N |
| Synonym | vanillic acid, acide vanillique, p-vanillic acid, benzoic acid, 4-hydroxy-3-methoxy, 3-methoxy-4-hydroxybenzoic acid, vanillate, m-anisic acid, 4-hydroxy, protocatechuic acid, 3-methyl ester, 4-hydroxy-3-methoxy-benzoic acid, unii-gm8q3jm2y8 |
| PubChem CID | 8468 |
| ChEBI | CHEBI:30816 |
| IUPAC Name | 4-hydroxy-3-methoxybenzoic acid |
| SMILES | COC1=CC(=CC=C1O)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC121350250
|
Thermo Scientific Chemicals
121350250 |
25 g | Glass bottle |
Each for $79.56
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 121-34-6 | |
| 168.15 | |
| WKOLLVMJNQIZCI-UHFFFAOYSA-N | |
| 8468 | |
| 4-hydroxy-3-methoxybenzoic acid |
| C8H8O4 | |
| MFCD00002551 | |
| vanillic acid, acide vanillique, p-vanillic acid, benzoic acid, 4-hydroxy-3-methoxy, 3-methoxy-4-hydroxybenzoic acid, vanillate, m-anisic acid, 4-hydroxy, protocatechuic acid, 3-methyl ester, 4-hydroxy-3-methoxy-benzoic acid, unii-gm8q3jm2y8 | |
| CHEBI:30816 | |
| COC1=CC(=CC=C1O)C(O)=O |
Specifications
| 121-34-6 | |
| 100.0 | |
| Brown to Colorless | |
| 97% min. (HPLC) | |
| C8H8O4 | |
| MFCD00002551 | |
| 10, 392 | |
| vanillic acid, acide vanillique, p-vanillic acid, benzoic acid, 4-hydroxy-3-methoxy, 3-methoxy-4-hydroxybenzoic acid, vanillate, m-anisic acid, 4-hydroxy, protocatechuic acid, 3-methyl ester, 4-hydroxy-3-methoxy-benzoic acid, unii-gm8q3jm2y8 | |
| WKOLLVMJNQIZCI-UHFFFAOYSA-N | |
| 4-hydroxy-3-methoxybenzoic acid | |
| 8468 | |
| 168.15 | |
| Liquid |
| 98.5 | |
| 210°C to 214°C | |
| Authentic | |
| Glass bottle | |
| HOC6H3(OCH3)CO2H | |
| 25 g | |
| 15, 10119 | |
| Solubility in water: .. Other solubilities: soluble in alcohol and ether | |
| COC1=CC(=CC=C1O)C(O)=O | |
| 168.15 | |
| CHEBI:30816 | |
| 97+% | |
| 4-Hydroxy-3-methoxybenzoic acid, 98.5+% |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 204-466-8
RTECSNumber : YW5300000
TSCA : TSCA
RUO – Research Use Only