missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Hydroxy-3,5-dimethylbenzonitrile 95.0+%, TCI America™
$32.72 - $121.28
Chemical Identifiers
| CAS | 4198-90-7 |
|---|---|
| Molecular Formula | C9H9NO |
| Molecular Weight (g/mol) | 147.177 |
| MDL Number | MFCD00221716 |
| InChI Key | WFYGXOWFEIOHCZ-UHFFFAOYSA-N |
| Synonym | 3,5-dimethyl-4-hydroxybenzonitrile, 4-cyano-2,6-dimethylphenol, 2,6-dimethyl-4-cyanophenol, 4-hydroxy-3,5-dimethyl-benzonitrile, 3,5-dimethyl-4-hydroxy benzonitrile, benzonitrile, 3,5-dimethyl-4-hydroxy, 4-hydroxy-3,5-dimethylbenzenecarbonitrile, benzonitrile, 4-hydroxy-3,5-dimethyl, pubchem11065, acmc-209jmm |
| PubChem CID | 20176 |
| IUPAC Name | 4-hydroxy-3,5-dimethylbenzonitrile |
| SMILES | CC1=CC(=CC(=C1O)C)C#N |
Chemical Identifiers
| 4198-90-7 | |
| 147.177 | |
| WFYGXOWFEIOHCZ-UHFFFAOYSA-N | |
| 20176 | |
| CC1=CC(=CC(=C1O)C)C#N |
| C9H9NO | |
| MFCD00221716 | |
| 3,5-dimethyl-4-hydroxybenzonitrile, 4-cyano-2,6-dimethylphenol, 2,6-dimethyl-4-cyanophenol, 4-hydroxy-3,5-dimethyl-benzonitrile, 3,5-dimethyl-4-hydroxy benzonitrile, benzonitrile, 3,5-dimethyl-4-hydroxy, 4-hydroxy-3,5-dimethylbenzenecarbonitrile, benzonitrile, 4-hydroxy-3,5-dimethyl, pubchem11065, acmc-209jmm | |
| 4-hydroxy-3,5-dimethylbenzonitrile |
Specifications
| 4198-90-7 | |
| C9H9NO | |
| 1 g | |
| WFYGXOWFEIOHCZ-UHFFFAOYSA-N | |
| 4-hydroxy-3,5-dimethylbenzonitrile | |
| 20176 | |
| ≥95.0% (GC) | |
| 4-Hydroxy-3,5-dimethylbenzonitrile |
| 125°C | |
| MFCD00221716 | |
| 3,5-dimethyl-4-hydroxybenzonitrile, 4-cyano-2,6-dimethylphenol, 2,6-dimethyl-4-cyanophenol, 4-hydroxy-3,5-dimethyl-benzonitrile, 3,5-dimethyl-4-hydroxy benzonitrile, benzonitrile, 3,5-dimethyl-4-hydroxy, 4-hydroxy-3,5-dimethylbenzenecarbonitrile, benzonitrile, 4-hydroxy-3,5-dimethyl, pubchem11065, acmc-209jmm | |
| CC1=CC(=CC(=C1O)C)C#N | |
| 147.177 | |
| 147.18 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : DI4359000
TSCA : No