missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3,5-dimethoxyphenylacetic Acid 98.0+%, TCI America™

Catalog No. p-7131065
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H14771G 1 g
H14775G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H14771G Supplier TCI America Supplier No. H14771G
Only null left

Chemical Identifiers

CAS 4385-56-2
Molecular Formula C10H12O5
Molecular Weight (g/mol) 212.201
MDL Number MFCD00016834
InChI Key BQBQKSSTFGCRQL-UHFFFAOYSA-N
PubChem CID 78093
IUPAC Name 2-(4-hydroxy-3,5-dimethoxyphenyl)acetic acid
SMILES COC1=CC(=CC(=C1O)OC)CC(=O)O

Specifications

Melting Point 130°C
Color White-Yellow
Quantity 1 g
Synonym 3,5-dimethoxy-4-hydroxyphenylacetic acid, homosyringic acid, 2-4-hydroxy-3,5-dimethoxyphenyl acetic acid, 4-hydroxy-3,5-dimethoxyphenyl acetic acid, 4-hydroxy-3,5-dimethoxyphenylacetic acid, benzeneacetic acid, 4-hydroxy-3,5-dimethoxy, acmc-1aewn, benzeneacetic acid,4-hydroxy-3,5-dimethoxy, 4-hydroxy-3,5-dimethoxy-phenyl-acetic acid, 4-hydroxy-3,5-dimethoxyphenyl acetic acid #
Formula Weight 212.20
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxy-3,5-dimethoxyphenylacetic Acid