missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3,5-diiodobenzoic acid, 97%

Catalog No. AAL0259509
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0259509 10 g
AAL0259518 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0259509 Supplier Thermo Scientific Chemicals Supplier No. L0259509
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 618-76-8 | C7H4I2O3 | 389.92 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 618-76-8
Molecular Formula C7H4I2O3
Molecular Weight (g/mol) 389.92
MDL Number MFCD00016532
InChI Key XREKTVACBXQCSB-UHFFFAOYSA-N
Synonym 3,5-diiodo-4-hydroxybenzoic acid, ioxynil acid, ioxynic acid, benzoic acid, 4-hydroxy-3,5-diiodo, unii-8c0qv5jk0d, 4-hydroxy-3,5-diiodo-benzoic acid, 8c0qv5jk0d, chembl83650, benzoic acid, 3,5-diiodo-4-hydroxy, acmc-1b674
PubChem CID 12065
IUPAC Name 4-hydroxy-3,5-diiodobenzoic acid
SMILES OC(=O)C1=CC(I)=C(O)C(I)=C1

Specifications

Melting Point ∽275°C (decomposition)
Quantity 10 g
Beilstein 2212145
Sensitivity Light sensitive
Formula Weight 389.91
Percent Purity 97%
Chemical Name or Material 4-Hydroxy-3,5-diiodobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-562-0
RTECSNumber DG8597500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only