missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-2-(trifluoromethyl)pyrimidine, 97%

Catalog No. AAL1311806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1311806 5 g
AAL1311803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1311806 Supplier Thermo Scientific Chemicals Supplier No. L1311806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1546-80-1 | C5H3F3N2O | 164.087 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1546-80-1
Molecular Formula C5H3F3N2O
Molecular Weight (g/mol) 164.087
MDL Number MFCD00151833
InChI Key PDCVDVCPQWFGAX-UHFFFAOYSA-N
Synonym 2-trifluoromethyl pyrimidin-4-ol, 4-hydroxy-2-trifluoromethyl pyrimidine, 2-trifluoromethyl-1h-pyrimidin-6-one, 2-trifluoromethyl-3h-pyrimidin-4-one, pubchem7199, acmc-1bux4, 2-trifluoromethyl-pyrimidin-4-ol, 2-trifluoromethyl-4-hydroxypyrimidine, 4-hydroxy-2-trifluoromethylpyrimidine, 4 3h-pyrimidinone,2-trifluoromethyl
PubChem CID 243570
IUPAC Name 2-(trifluoromethyl)-1H-pyrimidin-6-one
SMILES C1=CN=C(NC1=O)C(F)(F)F

Specifications

Melting Point 168°C to 171°C
Quantity 5 g
Beilstein 778887
Formula Weight 164.09
Percent Purity 97%
Chemical Name or Material 4-Hydroxy-2-(trifluoromethyl)pyrimidine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only