CAS RN 1546-80-1
IUPAC Name:
2-(trifluoromethyl)-3,4-dihydropyrimidin-4-one
Synonyms:
2-(trifluoromethyl)-3H-pyrimidin-4-one
Molecular Weight (g/mol):
164.09
Molecular Formula:
C5H3F3N2O
InChi Key:
PDCVDVCPQWFGAX-UHFFFAOYSA-N
SMILES:
FC(F)(F)C1=NC=CC(=O)N1
1
–
1
of
1
results
1
4-Hydroxy-2-(trifluoromethyl)pyrimidine, 97%
CAS: 1546-80-1 Molecular Formula: C5H3F3N2O Molecular Weight (g/mol): 164.087 MDL Number: MFCD00151833 InChI Key: PDCVDVCPQWFGAX-UHFFFAOYSA-N Synonym: 2-trifluoromethyl pyrimidin-4-ol,4-hydroxy-2-trifluoromethyl pyrimidine,2-trifluoromethyl-1h-pyrimidin-6-one,2-trifluoromethyl-3h-pyrimidin-4-one,pubchem7199,acmc-1bux4,2-trifluoromethyl-pyrimidin-4-ol,2-trifluoromethyl-4-hydroxypyrimidine,4-hydroxy-2-trifluoromethylpyrimidine,4 3h-pyrimidinone,2-trifluoromethyl PubChem CID: 243570 IUPAC Name: 2-(trifluoromethyl)-1H-pyrimidin-6-one SMILES: C1=CN=C(NC1=O)C(F)(F)F