missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-2-(trifluoromethyl)benzonitrile, 98%

Catalog No. AAH32158MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH32158MD 250 mg
AAH3215803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH32158MD Supplier Thermo Scientific Chemicals Supplier No. H32158MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 320-42-3 | C8H4F3NO | 187.121 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 320-42-3
Molecular Formula C8H4F3NO
Molecular Weight (g/mol) 187.121
MDL Number MFCD06797340
InChI Key MDBVIOUGHSKRMT-UHFFFAOYSA-N
Synonym 4-hydroxy-2-trifluoromethyl benzonitrile, 4-hydroxy-2-trifluoroemthyl benzonitrile, pubchem4807, 2-cyano-5-hydroxybenzotrifluoride, 4-cyano-3-trifluoromethyl phenol, 4-hydroxy-2-trifluoromethyl-benzonitrile, benzonitrile, 4-hydroxy-2-trifluoromethyl, 4-hydroxy-2-trifluoromethyl benzenecarbonitrile
PubChem CID 2783153
IUPAC Name 4-hydroxy-2-(trifluoromethyl)benzonitrile
SMILES C1=CC(=C(C=C1O)C(F)(F)F)C#N

Specifications

Melting Point 120°C to 124°C
Quantity 250 mg
Formula Weight 187.12
Percent Purity 98%
Chemical Name or Material 4-Hydroxy-2-(trifluoromethyl)benzonitrile
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only