missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-2-pyridinecarboxylic Acid 98.0+%, TCI America™

Catalog No. H14911G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H14911G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H14911G Supplier TCI America Supplier No. H14911G

Chemical Identifiers

CAS 22468-26-4
Molecular Formula C6H5NO3
Molecular Weight (g/mol) 139.11
MDL Number MFCD00521155,MFCD11656334
InChI Key MXXLHBCSVDDTIX-UHFFFAOYSA-N
Synonym 4-hydroxypicolinic acid, 4-hydroxypyridine-2-carboxylic acid, 4-hydroxy-2-pyridinecarboxylic acid, 4-oxo-1,4-dihydropyridine-2-carboxylic acid, 2-pyridinecarboxylic acid, 4-hydroxy, 4-hydroxypyridine-2-carboxylicacid, 2-pyridinecarboxylic acid, 1,4-dihydro-4-oxo, 4-hydroxy-picolinsaure, pubchem17131, acmc-209fwn
PubChem CID 735149
IUPAC Name 4-oxo-1,4-dihydropyridine-2-carboxylic acid
SMILES OC(=O)C1=CC(=O)C=CN1

Specifications

Melting Point 252°C
Color White-Yellow
Quantity 1 g
Formula Weight 139.11
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxy-2-pyridinecarboxylic Acid