missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-2'-nitrobiphenyl 97.0+%, TCI America™

Catalog No. H07535G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H07535G 5 g
H07531G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H07535G Supplier TCI America Supplier No. H07535G
Only null left

Chemical Identifiers

CAS 51264-59-6
Molecular Formula C12H9NO3
Molecular Weight (g/mol) 215.208
MDL Number MFCD00143326
InChI Key NRRCMTITSCFYOH-UHFFFAOYSA-N
PubChem CID 15541112
IUPAC Name 4-(2-nitrophenyl)phenol
SMILES C1=CC=C(C(=C1)C2=CC=C(C=C2)O)[N+](=O)[O-]

Specifications

Melting Point 115°C
Quantity 5 g
Synonym 2′-Nitrobiphenyl-4-ol, 4-(2-Nitrophenyl)phenol
Formula Weight 215.21
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxy-2′-nitrobiphenyl