CAS RN 51264-59-6
IUPAC Name:
2'-nitro-[1,1'-biphenyl]-4-ol
Synonyms:
2'-nitro-[1,1'-biphenyl]-4-ol
Molecular Weight (g/mol):
215.21
Molecular Formula:
C12H9NO3
InChi Key:
NRRCMTITSCFYOH-UHFFFAOYSA-N
SMILES:
OC1=CC=C(C=C1)C1=CC=CC=C1[N+]([O-])=O
1
–
1
of
1
results
1
4-Hydroxy-2'-nitrobiphenyl 97.0+%, TCI America™
CAS: 51264-59-6 Molecular Formula: C12H9NO3 Molecular Weight (g/mol): 215.208 MDL Number: MFCD00143326 InChI Key: NRRCMTITSCFYOH-UHFFFAOYSA-N PubChem CID: 15541112 IUPAC Name: 4-(2-nitrophenyl)phenol SMILES: C1=CC=C(C(=C1)C2=CC=C(C=C2)O)[N+](=O)[O-]