missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-2-methylquinoline, 98+%

Catalog No. p-7022012
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1458706 5 g
AAA1458714 25 g
AAA1458722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1458706 Supplier Thermo Scientific Chemicals Supplier No. A1458706

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 607-67-0 | C10H9NO | 159.19 g/mol

4-Hydroxy-2-methylquinoline is act as an intermediate in the synthesis of dequalinium chloride and as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxy-2-methylquinoline is act as an intermediate in the synthesis of dequalinium chloride and as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 607-67-0
Molecular Formula C10H9NO
Molecular Weight (g/mol) 159.19
MDL Number MFCD00006758,MFCD00518775
InChI Key NWINIEGDLHHNLH-UHFFFAOYSA-N
Synonym 2-methylquinolin-4-ol, 4-hydroxy-2-methylquinoline, 2-methyl-4-quinolinol, 4-quinolinol, 2-methyl, 4-hydroxyquinaldine, 2-methyl-4-hydroxyquinoline, 2-methyl-quinolin-4-ol, 2-methylquinolin-4 1h-one, 2-methyl-4 1h-quinolinone, 2-methyl-1,4-dihydroquinolin-4-one
PubChem CID 69089
SMILES CC1=CC(=O)C2=CC=CC=C2N1

Specifications

Melting Point 234°C to 236°C
Quantity 5 g
Beilstein 118320
Solubility Information Slightly soluble in water.
IUPAC Name 2-methyl-1H-quinolin-4-one
Formula Weight 159.19
Percent Purity ≥98%
Chemical Name or Material 4-Hydroxy-2-methylquinoline
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-140-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only