accessibility menu, dialog, popup

UserName

CAS RN 607-67-0

IUPAC Name: 2-methyl-1,4-dihydroquinolin-4-one
Synonyms: 2-methyl-1H-quinolin-4-one
Molecular Weight (g/mol): 159.19
Molecular Formula: C10H9NO
InChi Key: NWINIEGDLHHNLH-UHFFFAOYSA-N
SMILES: CC1=CC(=O)C2=CC=CC=C2N1
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)