missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-2-methyl-6-(trifluoromethyl)pyrimidine 98.0+%, TCI America™

Catalog No. H16021G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H16021G 1 g
H16025G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H16021G Supplier TCI America Supplier No. H16021G
Only null left

Chemical Identifiers

CAS 2836-44-4
Molecular Formula C6H5F3N2O
Molecular Weight (g/mol) 178.114
MDL Number MFCD03265321
InChI Key DBDRDWKBAGNHHS-UHFFFAOYSA-N
Synonym 2-methyl-6-trifluoromethyl pyrimidin-4-ol, 4-hydroxy-2-methyl-6-trifluoromethylpyrimidine, 4-hydroxy-2-methyl-6-trifluoromethyl pyrimidine, 4-pyrimidinol, 2-methyl-6-trifluoromethyl, 2-methyl-6-trifluoromethylpyrimidin-4-ol, 2-methyl-6-trifluoromethyl-4-pyrimidinol, 2-methyl-4-hydroxy-6-trifluoromethylpyrimidine, 2-methyl-6-trifluoromethyl-1h-pyrimidin-4-one, 2-methyl-6-trifluoromethyl pyrimidin-4 3h-one, 4 1h-pyrimidinone, 2-methyl-6-trifluoromethyl
PubChem CID 233901
IUPAC Name 2-methyl-6-(trifluoromethyl)-1H-pyrimidin-4-one
SMILES CC1=NC(=O)C=C(N1)C(F)(F)F

Specifications

Melting Point 143°C
Color White-Yellow
Quantity 1 g
Formula Weight 178.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxy-2-methyl-6-(trifluoromethyl)pyrimidine