missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-2-methoxybenzaldehyde 97.0+%, TCI America™

Catalog No. H11915G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H11915G 5 g
H11911G 1 g
H119125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H11915G Supplier TCI America Supplier No. H11915G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 18278-34-7
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00051964
InChI Key WBIZZNFQJPOKDK-UHFFFAOYSA-N
Synonym 4-hydroxy-o-anisaldehyde, benzaldehyde, 4-hydroxy-2-methoxy, 4-hydroxyl-2-methoxyl benzaldehyde, 4-formyl-3-methoxyphenol, 2-formyl-5-hydroxyanisole, 4-hydroxy-2-methoxybenaldehyde, 4-hydroxy-2-methoxybenzaldehdye, pubchem8130, acmc-209eju, argogel-mb-cho
PubChem CID 519541
IUPAC Name 4-hydroxy-2-methoxybenzaldehyde
SMILES COC1=CC(O)=CC=C1C=O

Specifications

Melting Point 158°C
Quantity 5 g
Formula Weight 152.15
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxy-2-methoxybenzaldehyde
EINECSNumber (3)-1200
TSCA No