accessibility menu, dialog, popup

UserName

CAS RN 18278-34-7

IUPAC Name: 4-hydroxy-2-methoxybenzaldehyde
Synonyms: 4-hydroxy-2-methoxybenzaldehyde
Molecular Weight (g/mol): 152.15
Molecular Formula: C8H8O3
InChi Key: WBIZZNFQJPOKDK-UHFFFAOYSA-N
SMILES: COC1=CC(O)=CC=C1C=O
Percent Purity 
  • (3)
  • (6)
Quantity 
  • (3)
  • (3)
  • (3)
Physical Form 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (6)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)