missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-2,2,6,6-tetramethylpiperidine 1-Oxyl Free Radical 98.0+%, TCI America™

Catalog No. p-7130771
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H08655G 5 g
H086525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H08655G Supplier TCI America Supplier No. H08655G

Chemical Identifiers

CAS 2226-96-2
Molecular Formula C9H18NO2
MDL Number MFCD00006478
Synonym 4-Hydroxy-TEMPO, TEMPOL

Specifications

Melting Point 72°C
Quantity 5 g
Formula Weight 172.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxy-2,2,6,6-tetramethylpiperidine 1-Oxyl Free Radical
EINECSNumber (5)-6249
RTECSNumber TN8991000
TSCA Yes