missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Hydroxy-1-methyl-2-quinolone 98.0+%, TCI America™
Supplier: TCI America M100925G
Specifications
| 4-Hydroxy-1-methyl-2-quinolone | |
| 270°C | |
| MFCD00024052 | |
| 4-hydroxy-1-methyl-2-quinolone, 4-hydroxy-1-methylcarbostyril, 2 1h-quinolinone, 4-hydroxy-1-methyl, 4-hydroxy-1-methyl-2 1h-quinolone, 4-hydroxy-n-methylcarbostyril, 1-methyl-4-hydroxycarbostyril, 4-hydroxy-1-methylquinolin-2 1h-one, 1-methyl-4-hydroxy-2-chinolon, 4-hydroxy-1-methyl-2 1h-quinolinone, carbostyril, 4-hydroxy-1-methyl | |
| CN1C(O)=CC(=O)C2=CC=CC=C12 | |
| 175.19 | |
| 175.19 | |
| Crystalline Powder |
| 1677-46-9 | |
| C10H9NO2 | |
| 25 g | |
| VWRAQWACTKTNAO-UHFFFAOYSA-N | |
| 2-hydroxy-1-methyl-1,4-dihydroquinolin-4-one | |
| 54686436 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 1677-46-9 | |
| 175.19 | |
| VWRAQWACTKTNAO-UHFFFAOYSA-N | |
| 54686436 | |
| CN1C(O)=CC(=O)C2=CC=CC=C12 |
| C10H9NO2 | |
| MFCD00024052 | |
| 4-hydroxy-1-methyl-2-quinolone, 4-hydroxy-1-methylcarbostyril, 2 1h-quinolinone, 4-hydroxy-1-methyl, 4-hydroxy-1-methyl-2 1h-quinolone, 4-hydroxy-n-methylcarbostyril, 1-methyl-4-hydroxycarbostyril, 4-hydroxy-1-methylquinolin-2 1h-one, 1-methyl-4-hydroxy-2-chinolon, 4-hydroxy-1-methyl-2 1h-quinolinone, carbostyril, 4-hydroxy-1-methyl | |
| 2-hydroxy-1-methyl-1,4-dihydroquinolin-4-one |
Safety and Handling
RTECSNumber : FG7350000
TSCA : Yes