missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-1-methyl-2-quinolone 98.0+%, TCI America™

Catalog No. p-7132495
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M10095G 5 g
M100925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M10095G Supplier TCI America Supplier No. M10095G
Only null left

Chemical Identifiers

CAS 1677-46-9
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.19
MDL Number MFCD00024052
InChI Key VWRAQWACTKTNAO-UHFFFAOYSA-N
Synonym 4-hydroxy-1-methyl-2-quinolone, 4-hydroxy-1-methylcarbostyril, 2 1h-quinolinone, 4-hydroxy-1-methyl, 4-hydroxy-1-methyl-2 1h-quinolone, 4-hydroxy-n-methylcarbostyril, 1-methyl-4-hydroxycarbostyril, 4-hydroxy-1-methylquinolin-2 1h-one, 1-methyl-4-hydroxy-2-chinolon, 4-hydroxy-1-methyl-2 1h-quinolinone, carbostyril, 4-hydroxy-1-methyl
PubChem CID 54686436
IUPAC Name 2-hydroxy-1-methyl-1,4-dihydroquinolin-4-one
SMILES CN1C(O)=CC(=O)C2=CC=CC=C12

Specifications

Melting Point 270°C
Quantity 5 g
Formula Weight 175.19
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxy-1-methyl-2-quinolone
RTECSNumber FG7350000
TSCA Yes