missing translation for 'onlineSavingsMsg'
Learn More

4-Hydrazinobenzonitrile Hydrochloride 98.0+%, TCI America™

Catalog No. p-7131038
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H14255G 5 g
H142525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H14255G Supplier TCI America Supplier No. H14255G
Only null left

Chemical Identifiers

CAS 2863-98-1
Molecular Formula C7H7N3
Molecular Weight (g/mol) 133.15
MDL Number MFCD00673994
InChI Key DZUUSHCOMPROCJ-UHFFFAOYSA-N
Synonym 4-cyanophenylhydrazine hydrochloride, 4-hydrazinylbenzonitrile hydrochloride, 4-hydrazinobenzonitrile hydrochloride, 4-cyanophenylhydrazine hcl, benzonitrile, 4-hydrazino-, monohydrochloride, 4-hydrazino-benzonitrile; hydrochloride, pubchem13684, 4-cyanophenylhydrazine-hcl, acmc-1co7p, ksc205o1j
PubChem CID 16212962
IUPAC Name 4-hydrazinylbenzonitrile
SMILES NNC1=CC=C(C=C1)C#N

Specifications

Melting Point 243°C
Quantity 5 g
Formula Weight 169.61
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydrazinobenzonitrile Hydrochloride