missing translation for 'onlineSavingsMsg'
Learn More

4-Hydrazinobenzenesulfonic Acid Hemihydrate 98.0+%, TCI America™

Catalog No. H017625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H017625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H017625G Supplier TCI America Supplier No. H017625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 854689-07-9
Molecular Formula C6H10N2O4S
Molecular Weight (g/mol) 206.216
MDL Number MFCD00480958
InChI Key OSOMMFVBTYFMBI-UHFFFAOYSA-N
Synonym 4-hydrazinylbenzenesulfonic acid hydrate, 4-hydrazinobenzenesulfonic acid hydrate, phenylhydrazine-4-sulfonic acid hemihydrate, benzenesulfonic acid, 4-hydrazino-, monohydrate 9ci
PubChem CID 2849446
IUPAC Name 4-hydrazinylbenzenesulfonic acid;hydrate
SMILES C1=CC(=CC=C1NN)S(=O)(=O)O.O

Specifications

Quantity 25 g
Formula Weight 188.20
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydrazinobenzenesulfonic Acid Hemihydrate
EINECSNumber (3)-1997
RTECSNumber DB6900000
TSCA Yes