missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 4-HNE Competitive ELISA Kit

Catalog No. EEL163
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EEL163 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EEL163 Supplier Invitrogen™ Supplier No. EEL163

ELISA

The 4 Hydroxynonenal (4 HNE) ELISA quantitates 4 HNE in serum, plasma and other biological fluids. Principle of the method The Competitive multispecies ELISA research use only kit is designed to quantitatively measure The 4 Hydroxynonenal (4 HNE) independent of species. A 4 Hydroxynonenal standard is provided to generate a standard curve for the assay and all samples are read off a user generated standard curve.Standards or diluted samples are pipetted into a coated microtiter plate and Biotinylated Detection Ab specific to 4 Hydroxynonenal is added. Excess conjugate and unbound sample or standard are washed from the plate, and Avidin conjugated to Horseradish Peroxidase (HRP) is added to each well and incubated. Then a TMB substrate solution is added to each well. The enzyme substrate reaction is terminated by the addition of stop solution and the color change is measured on a microplate reader. The concentration of 4 Hydroxynonenal in the samples is then determined by comparing the OD of the samples to the standard curve. Rigorous validation: Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation. For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

4-Hydroxynonenal, or 4-hydroxy-2-nonenal or 4-HNE or HNE, is an alpha, beta-unsaturated hydroxyalkenal that is produced by lipid peroxidation in cells. 4-HNE is the primary alpha, beta-unsaturated hydroxyalkenal formed in this process.

Specifications

Assay Range 0.63-40 ng/mL
Assay Sensitivity 0.38 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Cell Lysates, Plasma, Serum, Tissue Homogenates
Target Species All Species
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias 4HNE; 4-HNE; 4-hydroxy Hexenal; 4-Hydroxy Nonenal; 4-hydroxy-2-nonenal; 4-Hydroxynonenal; HNE4HNE; 4-HNE; 4-hydroxy Hexenal; 4-Hydroxy Nonenal; 4-hydroxy-2-nonenal; 4-Hydroxynonenal; HNE;
Interassay CV <10%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Biotinylated Detection Antibody, HRP Conjugate, Standard & Sample Diluent, Biotinylated Detection Antibody Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer, Standard, Biotinylated Detection Antibody, HRP Conjugate, Standard & Sample Diluent, Biotinylated Detection Antibody Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer
Label or Dye Biotin
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Cell Lysate 50 μL, Plasma 50 μL, Serum 50 μL, Tissue Homogenate 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Refer to product documentation for component specific storage temperature.
Target 4-Hydroxynonenal
Test Time 1 hr 20 min
Total Assay Time 2 hr 30 min
Show More Show Less