missing translation for 'onlineSavingsMsg'
Learn More

4-Hexyloxyphthalonitrile 95.0+%, TCI America™

Catalog No. p-7130675
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H06911G 1 g
H06915G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H06911G Supplier TCI America Supplier No. H06911G
Only null left

Chemical Identifiers

CAS 104949-82-8
Molecular Formula C14H16N2O
Molecular Weight (g/mol) 228.295
MDL Number MFCD00059077
InChI Key BMXSATBWGFIAPA-UHFFFAOYSA-N
Synonym 1,2-Dicyano-4-hexyloxybenzene
PubChem CID 538284
IUPAC Name 4-hexoxybenzene-1,2-dicarbonitrile
SMILES CCCCCCOC1=CC(=C(C=C1)C#N)C#N

Specifications

Color Yellow
Quantity 1 g
Formula Weight 228.30
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Hexyloxyphthalonitrile