missing translation for 'onlineSavingsMsg'
Learn More

4-Hexylbenzoic Acid 98.0+%, TCI America™

Catalog No. H08295G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H08295G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H08295G Supplier TCI America Supplier No. H08295G

Chemical Identifiers

CAS 21643-38-9
Molecular Formula C13H18O2
Molecular Weight (g/mol) 206.285
MDL Number MFCD00013997
InChI Key CPEPWESLFZVUEP-UHFFFAOYSA-N
Synonym benzoic acid, 4-hexyl, 4-n-hexylbenzoic acid, p-hexylbenzoic acid, benzoic acid, p-hexyl, p-n-hexylbenzoic acid, 4n-hexylbenzoic acid, maybridge1_002028, para-n-hexyl benzoic acid, acmc-1chu5, ksc201s3p
PubChem CID 30725
IUPAC Name 4-hexylbenzoic acid
SMILES CCCCCCC1=CC=C(C=C1)C(=O)O

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 206.29
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hexylbenzoic Acid
EINECSNumber (3)-1338
RTECSNumber DH1485000
TSCA Yes