missing translation for 'onlineSavingsMsg'
Learn More

4-Hexyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene 98.0+%, TCI America™

Catalog No. H12945G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H12945G 5 g
H12941G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H12945G Supplier TCI America Supplier No. H12945G

Chemical Identifiers

CAS 883742-29-8
Molecular Formula C16H27BO2S
Molecular Weight (g/mol) 294.26
MDL Number MFCD14708172
InChI Key QEFQFMZHWUCJOA-UHFFFAOYSA-N
Synonym 2-(4-Hexyl-2-thienyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-Hexyl-2-thiopheneboronic Acid Pinacol Ester
PubChem CID 44630029
IUPAC Name 2-(4-hexylthiophen-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CCCCCCC1=CSC(=C1)B1OC(C)(C)C(C)(C)O1

Specifications

Color Yellow
Quantity 5 g
Formula Weight 294.26
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Hexyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene
TSCA No
Recommended Storage Refrigerator