CAS RN 883742-29-8
IUPAC Name:
2-(4-hexylthiophen-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms:
2-(4-hexylthiophen-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol):
294.26
Molecular Formula:
C16H27BO2S
InChi Key:
QEFQFMZHWUCJOA-UHFFFAOYSA-N
SMILES:
CCCCCCC1=CSC(=C1)B1OC(C)(C)C(C)(C)O1
1
–
1
of
1
results
1
4-Hexyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene 98.0+%, TCI America™
CAS: 883742-29-8 Molecular Formula: C16H27BO2S Molecular Weight (g/mol): 294.26 MDL Number: MFCD14708172 InChI Key: QEFQFMZHWUCJOA-UHFFFAOYSA-N Synonym: 2-(4-Hexyl-2-thienyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-Hexyl-2-thiopheneboronic Acid Pinacol Ester PubChem CID: 44630029 IUPAC Name: 2-(4-hexylthiophen-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane SMILES: CCCCCCC1=CSC(=C1)B1OC(C)(C)C(C)(C)O1