missing translation for 'onlineSavingsMsg'
Learn More

4-(Heptyloxy)benzoic Acid 98.0+%, TCI America™

Catalog No. H024825G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H024825G 25 g
H0248500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H024825G Supplier TCI America Supplier No. H024825G
Only null left

Chemical Identifiers

CAS 15872-42-1
Molecular Formula C14H20O3
Molecular Weight (g/mol) 236.31
MDL Number MFCD00013992
InChI Key ZRVIYEJYXIDATJ-UHFFFAOYSA-N
Synonym 4-heptyloxy benzoic acid, 4-heptyloxybenzoic acid, 4-n-heptyloxybenzoic acid, p-heptoxybenzoic acid, p-heptyloxybenzoic acid, benzoic acid, 4-heptyloxy, unii-bj0yzc8zy8, benzoic acid, p-heptyloxy, p-n-heptyloxybenzoic acid, bj0yzc8zy8
PubChem CID 85154
ChEBI CHEBI:34405
IUPAC Name 4-(heptyloxy)benzoic acid
SMILES CCCCCCCOC1=CC=C(C=C1)C(O)=O

Specifications

Melting Point 145°C
Color White
Quantity 25 g
Formula Weight 236.31
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Heptyloxy)benzoic Acid