missing translation for 'onlineSavingsMsg'
Learn More

4-Heptylamine 98.0+%, TCI America™

Catalog No. p-7130651
Change view
Click to view available options
Quantity:
10 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H063710ML 10 mL
H063725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H063710ML Supplier TCI America Supplier No. H063710ML

Chemical Identifiers

CAS 16751-59-0
Molecular Formula C7H17N
Molecular Weight (g/mol) 115.22
MDL Number MFCD00014814
InChI Key CLJMMQGDJNYDER-UHFFFAOYSA-N
Synonym 4-Aminoheptane
PubChem CID 38665
IUPAC Name heptan-4-amine
SMILES CCCC(CCC)N

Specifications

Color Colorless
Boiling Point 140°C
Quantity 10 mL
UN Number 2733
Formula Weight 115.22
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Heptylamine
EINECSNumber (2)-0133
RTECSNumber EO6025000
TSCA Yes