accessibility menu, dialog, popup

UserName

H3CNH2CH3

CAS RN 16751-59-0

IUPAC Name: heptan-4-amine
Synonyms: 4-heptanamine
Molecular Weight (g/mol): 115.22
Molecular Formula: C7H17N
InChi Key: CLJMMQGDJNYDER-UHFFFAOYSA-N
SMILES: CCCC(N)CCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results