missing translation for 'onlineSavingsMsg'
Learn More

4-Heptanol 97.0+%, TCI America™

Catalog No. H003625ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H003625ML 25 mL
H0036100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H003625ML Supplier TCI America Supplier No. H003625ML

Chemical Identifiers

CAS 589-55-9
Molecular Formula C7H16O
Molecular Weight (g/mol) 116.204
MDL Number MFCD00021934
InChI Key YVBCULSIZWMTFY-UHFFFAOYSA-N
Synonym 4-heptanol, dipropylcarbinol, unii-yg7b8091bp, di-n-propylcarbinol, heptanol-4, 4-heptyl alcohol, n-heptan-4-ol, acmc-209m8k, yvbculsizwmtfy-uhfffaoysa, ch3 ch2 2choh ch2 2ch3
PubChem CID 11513
IUPAC Name heptan-4-ol
SMILES CCCC(CCC)O

Specifications

Melting Point -42°C
Color Colorless
Boiling Point 155°C
Quantity 25 mL
UN Number 1987
Formula Weight 116.20
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Heptanol
EINECSNumber (2)-0217
TSCA Yes