accessibility menu, dialog, popup

UserName

CAS RN 589-55-9

IUPAC Name: heptan-4-ol
Synonyms: 4-heptanol
Molecular Weight (g/mol): 116.2
Molecular Formula: C7H16O
InChi Key: YVBCULSIZWMTFY-UHFFFAOYSA-N
SMILES: CCCC(O)CCC
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)