missing translation for 'onlineSavingsMsg'
Learn More

4-Glycidyloxycarbazole 98.0+%, TCI America™

Catalog No. G02965G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G02965G 5 g
G029625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G02965G Supplier TCI America Supplier No. G02965G

Chemical Identifiers

CAS 51997-51-4
Molecular Formula C15H13NO2
Molecular Weight (g/mol) 239.274
MDL Number MFCD03411884
InChI Key SVWKIGRDISDRLO-UHFFFAOYSA-N
Synonym 4-(2,3-Epoxypropoxy)carbazole
PubChem CID 5020710
IUPAC Name 4-(oxiran-2-ylmethoxy)-9H-carbazole
SMILES C1C(O1)COC2=CC=CC3=C2C4=CC=CC=C4N3

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 239.27
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4-Glycidyloxycarbazole