CAS RN 51997-51-4
IUPAC Name:
4-[(oxiran-2-yl)methoxy]-9H-carbazole
Synonyms:
4-(oxiran-2-ylmethoxy)-9H-carbazole
Molecular Weight (g/mol):
239.27
Molecular Formula:
C15H13NO2
InChi Key:
SVWKIGRDISDRLO-UHFFFAOYNA-N
SMILES:
C(OC1=CC=CC2=C1C1=C(N2)C=CC=C1)C1CO1
1
–
1
of
1
results
1
4-Glycidyloxycarbazole 98.0+%, TCI America™
CAS: 51997-51-4 Molecular Formula: C15H13NO2 Molecular Weight (g/mol): 239.274 MDL Number: MFCD03411884 InChI Key: SVWKIGRDISDRLO-UHFFFAOYSA-N Synonym: 4-(2,3-Epoxypropoxy)carbazole PubChem CID: 5020710 IUPAC Name: 4-(oxiran-2-ylmethoxy)-9H-carbazole SMILES: C1C(O1)COC2=CC=CC3=C2C4=CC=CC=C4N3