missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ 4 ft. Logic™ Class II, Type A2 Biosafety Cabinet, US Model

Logic Class II, Type A2 Biosafety Cabinets offer personnel, product and environmental protection from hazardous particulates requiring BSL 1-4 containment. All models ensure the highest standard of safety and are listed to NSF/ANSI 49. Logic cabinets maximize safety, performance, and comfort.

$25,163.88 - $28,238.22

Specifications

Description 4 ft. Logic Class II, Type A2 Biosafety Cabinet
Product Type Class II, Type A2 Bio Safety Cabinet
View More Specs

Products 8
Catalog Number Mfr. No. Includes Plug Type Sash Opening (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Includes Plug Type Sash Opening (Metric) Price Quantity & Availability  
01812621
View Documents Promotion Details
Labconco™
302480002
Cabinet North America (115 V, 15 A) 20.3 cm N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
01812619
View Documents Promotion Details
Labconco™
302480102
Cabinet, Base Stand North America (115 V, 15 A) 20.3 cm N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
01812617
View Documents Promotion Details
Labconco™
302481002
Cabinet, Accessory Package North America (115 V, 15 A) 20.3 cm N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
01812615
View Documents Promotion Details
Labconco™
302481102
Cabinet, Accessory Package, Base Stand North America (115 V, 15 A) 20.3 cm N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
01812637
View Documents Promotion Details
Labconco™
302410002
Cabinet North America (115 V, 15 A) 25 cm N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
01812635
View Documents Promotion Details
Labconco™
302410102
Cabinet, Base Stand North America (115 V, 15 A) 25 cm N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
01812633
View Documents Promotion Details
Labconco™
302411002
Cabinet, Accessory Package North America (115 V, 15 A) 25 cm N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
01812631
View Documents Promotion Details
Labconco™
302411102
Cabinet, Accessory Package, Base Stand North America (115 V, 15 A) 25 cm N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Performance Features:

  • Safe Class II airflows (approx. 70% recirculating, 30% exhausted) that provide personnel, product, and environmental protection. Cabinet interior provides ISO Class 5 (Grade A) or better working conditions
  • Constant Airflow Profile™ (CAP) and BalanceFlow™ technology maintain safe cabinet airflows – even as HEPA filters load. Optimized airflow minimizes energy use and noise output to industry-leading levels
  • Logic OS with 5 in. line-of-sight touchscreen display. Access cabinet information such as filter life, alerts and alarms, data logs. Remote connectivity, particle counter integration, and certification records elevate user control of the cabinet
  • Smart-Start™ and Night-Smart™ modes simplify startup and shut-down procedures to help ensure consistency and cleanliness before, during, and after work
  • Inclination Technology™ improves ergonomic working conditions by maximizing comfort and accessibility during long periods of work
  • Mag-Zone™ area and mating accessories keep common items such as pipettes and biohazard waste organized and within reach
  • Full, six-year warranty on all Class II biosafety cabinets
  • Optional Configurations
  • Accessory Package: UV lamp (254 nm), right-side service fixture with ball-type valve and right and left-side Vacu-Pass™ Cord and Cable Portals
  • Telescoping base stand with shelf and leveling feet
Specifications

Specifications

4 ft. Logic Class II, Type A2 Biosafety Cabinet
Class II, Type A2 Bio Safety Cabinet
Videos
SDS
Documents

Documents