missing translation for 'onlineSavingsMsg'
Learn More

4-Formylphenylboronic acid pinacol ester, 97%

Catalog No. AC445040250
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC445040250 25 g
AC445040050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC445040250 Supplier Thermo Scientific Chemicals Supplier No. 445040250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 128376-64-7 | C13H17BO3 | 232.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 128376-64-7
Molecular Formula C13H17BO3
Molecular Weight (g/mol) 232.09
MDL Number MFCD04972375
InChI Key DMBMXJJGPXADPO-UHFFFAOYSA-N
Synonym 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzaldehyde, 4-formylphenylboronic acid pinacol ester, 4-formylbenzeneboronic acid, pinacol ester, 4-formylphenylboronic acid pinacolate, 4-formylphenylboronic acid pinacol cyclic ester, 4-formylphenylboronic acid, pinacol ester, 4-tetramethyl-1,3,2-dioxaborolan-2-yl benzaldehyde, 4-formylbenzeneboronic acid pinacol ester, benzaldehyde, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-formylphenylboronic acid, pinacol cyclic ester
PubChem CID 2769536
IUPAC Name 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde
SMILES CC1(C)OB(OC1(C)C)C1=CC=C(C=O)C=C1

Specifications

Melting Point 59.0°C to 63.0°C
Assay Percent Range 96% min. (GC)
Quantity 25 g
Formula Weight 232.09
Percent Purity 97%
Chemical Name or Material 4-Formylphenylboronic acid pinacol ester
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only