missing translation for 'onlineSavingsMsg'
Learn More

4-Formylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. p-7129690
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F04461G 1 g
F04465G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F04461G Supplier TCI America Supplier No. F04461G
Only null left

Chemical Identifiers

CAS 87199-17-5
Molecular Formula C7H7BO3
Molecular Weight (g/mol) 149.94
MDL Number MFCD00151823
InChI Key VXWBQOJISHAKKM-UHFFFAOYSA-N
Synonym 4-formylphenyl boronic acid, 4-formylbenzeneboronic acid, 4-boronobenzaldehyde, boronic acid, 4-formylphenyl, boronic acid, b-4-formylphenyl, p-formylphenylboronic acid, 4-dihydroxyboryl benzaldehyde, 4-dihydroxyboranyl benzaldehyde, 4-benzaldehyde boronic acid, 4-formyl-phenylboronic acid
PubChem CID 591073
IUPAC Name (4-formylphenyl)boronic acid
SMILES OB(O)C1=CC=C(C=O)C=C1

Specifications

Melting Point 242°C
Color White-Yellow
Quantity 1 g
Formula Weight 149.94
Physical Form Crystalline Powder
Chemical Name or Material 4-Formylphenylboronic Acid (contains varying amounts of Anhydride)
EINECSNumber (3)-4642
TSCA Yes