missing translation for 'onlineSavingsMsg'
Learn More

4-Formylbenzonitrile 98.0+%, TCI America™

Catalog No. C04435G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C04435G 5 g
C044325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C04435G Supplier TCI America Supplier No. C04435G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 105-07-7
Molecular Formula C8H5NO
Molecular Weight (g/mol) 131.13
MDL Number MFCD00003376
InChI Key WZWIQYMTQZCSKI-UHFFFAOYSA-N
Synonym 4-cyanobenzaldehyde, benzonitrile, 4-formyl, p-cyanobenzaldehyde, p-formylbenzonitrile, benzaldehyde, p-cyano, benzonitrile, p-formyl, usaf kf-1, terephthalaldehydonitrile, p-cyanobenzenecarboxaldehyde, 4-formyl-benzonitrile
PubChem CID 66042
IUPAC Name 4-formylbenzonitrile
SMILES O=CC1=CC=C(C=C1)C#N

Specifications

Melting Point 100°C
Color White-Yellow
Boiling Point 133°C
Quantity 5 g
Formula Weight 131.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Formylbenzonitrile
RTECSNumber CU5250000
TSCA Yes