missing translation for 'onlineSavingsMsg'
Learn More

4'-Formylbenzo-18-crown 6-Ether 98.0+%, TCI America™

Catalog No. f0451500mg
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0451500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F0451500MG Supplier TCI America Supplier No. F0451500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 60835-74-7
Molecular Formula C17H24O7
Molecular Weight (g/mol) 340.372
MDL Number MFCD00388882
InChI Key ALMXRNQJWRGNMG-UHFFFAOYSA-N
Synonym 2,3-(4-Formylbenzo)-1,4,7,10,13,16-hexaoxacyclooctadec-2-ene
PubChem CID 12342344
IUPAC Name 2,5,8,11,14,17-hexaoxabicyclo[16.4.0]docosa-1(18),19,21-triene-20-carbaldehyde
SMILES C1COCCOCCOC2=C(C=CC(=C2)C=O)OCCOCCO1

Specifications

Quantity 500 mg
Formula Weight 340.37
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Formylbenzo-18-crown 6-Ether