missing translation for 'onlineSavingsMsg'
Learn More

4-Formylbenzeneboronic acid pinacol ester, 98%

Catalog No. AAH6419203
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6419203 1 g
AAH6419206 5 g
AAH6419214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6419203 Supplier Thermo Scientific Chemicals Supplier No. H6419203

CAS: 128376-64-7 | C13H17BO3 | 232.09 g/mol

A boronic acid benzaldehyde compound for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A boronic acid benzaldehyde compound for proteomics research.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 128376-64-7
Molecular Formula C13H17BO3
Molecular Weight (g/mol) 232.09
MDL Number MFCD04972375
InChI Key DMBMXJJGPXADPO-UHFFFAOYSA-N
Synonym 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzaldehyde, 4-formylphenylboronic acid pinacol ester, 4-formylbenzeneboronic acid, pinacol ester, 4-formylphenylboronic acid pinacolate, 4-formylphenylboronic acid pinacol cyclic ester, 4-formylphenylboronic acid, pinacol ester, 4-tetramethyl-1,3,2-dioxaborolan-2-yl benzaldehyde, 4-formylbenzeneboronic acid pinacol ester, benzaldehyde, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-formylphenylboronic acid, pinacol cyclic ester
PubChem CID 2769536
IUPAC Name 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde
SMILES CC1(C)OB(OC1(C)C)C1=CC=C(C=O)C=C1

Specifications

Melting Point 54°C to 59°C
Color White
Quantity 1 g
Beilstein 7704448
Solubility Information Slightly soluble in water.
Formula Weight 232.09
Percent Purity 98%
Physical Form Crystalline Solid
Chemical Name or Material 4-Formylbenzeneboronic acid pinacol ester
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only