missing translation for 'onlineSavingsMsg'
Learn More

4-Formylbenzeneboronic acid, 97%

Catalog No. AAB2519914
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2519914 25 g
AAB2519903 1 g
AAB2519906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2519914 Supplier Thermo Scientific Chemicals Supplier No. B2519914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 87199-17-5 | C7H7BO3 | 149.94 g/mol

4-Formylbenzeneboronic acid acts as a reagent used for Palladium-catalyzed arylation Suzuki-Miyaura cross-coupling in water, copper-mediated ligandless aerobic fluoroalkylation of arylboronic acids with fluoroalkyl iodides, ligand-free copper-catalyzed coupling of nitro arenes with arylboronic acids, triethylamine-catalyzed three-component Hantzsch condensations, copper-catalyzed nitrations, oxidative mono-cleavage of dialkenes catalyzed by Trametes hirsuta, palladacycle-catalyzed cross-coupling of arylboronic acids with carboxylic anhydrides or acyl chlorides, palladium-catalyzed aerobic oxidative cross-coupling reactions. It acts as a reagent used in preparation of sensitizers with dithiafulvenyl unit as electron donor for high-efficiency dye-sensitized solar cells, a novel protein synthesis inhibitor active against Gram-positive bacteria.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Formylbenzeneboronic acid acts as a reagent used for Palladium-catalyzed arylation Suzuki-Miyaura cross-coupling in water, copper-mediated ligandless aerobic fluoroalkylation of arylboronic acids with fluoroalkyl iodides, ligand-free copper-catalyzed coupling of nitro arenes with arylboronic acids, triethylamine-catalyzed three-component Hantzsch condensations, copper-catalyzed nitrations, oxidative mono-cleavage of dialkenes catalyzed by Trametes hirsuta, palladacycle-catalyzed cross-coupling of arylboronic acids with carboxylic anhydrides or acyl chlorides, palladium-catalyzed aerobic oxidative cross-coupling reactions. It acts as a reagent used in preparation of sensitizers with dithiafulvenyl unit as electron donor for high-efficiency dye-sensitized solar cells, a novel protein synthesis inhibitor active against Gram-positive bacteria.

Solubility
Slightly Soluble in water.

Notes
Air Sensitive. Store away from air and oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 87199-17-5
Molecular Formula C7H7BO3
Molecular Weight (g/mol) 149.94
MDL Number MFCD00151823
InChI Key VXWBQOJISHAKKM-UHFFFAOYSA-N
Synonym 4-formylphenyl boronic acid, 4-formylbenzeneboronic acid, 4-boronobenzaldehyde, boronic acid, 4-formylphenyl, boronic acid, b-4-formylphenyl, p-formylphenylboronic acid, 4-dihydroxyboryl benzaldehyde, 4-dihydroxyboranyl benzaldehyde, 4-benzaldehyde boronic acid, 4-formyl-phenylboronic acid
PubChem CID 591073
IUPAC Name (4-formylphenyl)boronic acid
SMILES OB(O)C1=CC=C(C=O)C=C1

Specifications

Melting Point 260°C to 266°C
Quantity 25 g
Beilstein 3030770
Sensitivity Air Sensitive
Solubility Information Slightly Soluble in water.
Formula Weight 149.94
Percent Purity 97%
Chemical Name or Material 4-Formylbenzeneboronic acid
Hazard Category H317
Hazard Statement GHS H Statement
H317
May cause an allergic skin reaction.
Precautionary Statement P261-P272-P280g-P302+P352-P333+P313-P363-P501c
EINECSNumber 438-670-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only