missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorotoluene 99.0+%, TCI America™

Catalog No. F0041500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0041500ML 500 mL
F004125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F0041500ML Supplier TCI America Supplier No. F0041500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 352-32-9
Molecular Formula C7H7F
Molecular Weight (g/mol) 110.131
MDL Number MFCD00000358
InChI Key WRWPPGUCZBJXKX-UHFFFAOYSA-N
Synonym 4-fluorotoluene, p-fluorotoluene, benzene, 1-fluoro-4-methyl, p-fluoromethylbenzene, toluene, p-fluoro, 1-methyl-4-fluorobenzene, p-fluoromethyl benzene, unii-nud6wz1b8f, p-tolyl fluoride, para-fluorotoluene
PubChem CID 9603
IUPAC Name 1-fluoro-4-methylbenzene
SMILES CC1=CC=C(C=C1)F

Specifications

Melting Point -56°C
Color Yellow
Boiling Point 116°C
Quantity 500 mL
UN Number 2388
Formula Weight 110.13
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Fluorotoluene
EINECSNumber (3)-0083
RTECSNumber XT2580000
TSCA Yes