missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorothiophenol, 97%

Catalog No. AAA1760006
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1760006 5 g
AAA1760014 25 g
AAA1760022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1760006 Supplier Thermo Scientific Chemicals Supplier No. A1760006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 371-42-6 | C6H5FS | 128.164 g/mol

It is used in the synthesis of Bicalutamide, an anti-cancer drug.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the synthesis of Bicalutamide, an anti-cancer drug.

Solubility
Not miscible or difficult to mix in water.

Notes
Air Sensitive. Keep away from oxidizing agents and air. Store under inert gas.

Chemical Identifiers

CAS 371-42-6
Molecular Formula C6H5FS
Molecular Weight (g/mol) 128.164
MDL Number MFCD00004846
InChI Key OKIHXNKYYGUVTE-UHFFFAOYSA-N
Synonym 4-fluorothiophenol, p-fluorothiophenol, 4-fluorobenzene-1-thiol, 4-fluoro thiophenol, p-fluorophenyl mercaptan, benzenethiol, 4-fluoro, 4-fluoro-benzenethiol, 4-mercaptofluorobenzene, 4-fluorthiophenol, p-fluoro-thiophenol
PubChem CID 67789
IUPAC Name 4-fluorobenzenethiol
SMILES C1=CC(=CC=C1F)S

Specifications

Density 1.203
Boiling Point 170°C to 172°C
Flash Point 57°C (135°F)
Odor Unpleasant
Refractive Index 1.55
Quantity 5 g
UN Number UN3071
Beilstein 1446350
Sensitivity Air Sensitive
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 128.17
Percent Purity ≥98%
Assay (dry wt.), water <7%
Chemical Name or Material 4-Fluorothiophenol
Show More Show Less
Hazard Category H226-H301+H311+H331-H315-H319-H335
Hazard Statement GHS H Statement
H300-H310-H331-H226-H315-H319-H317-H335
Fatal if swallowed.
Fatal in contact with skin.
Toxic if inhaled.
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: III
EINECSNumber 206-737-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only