missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorothioanisole 98.0+%, TCI America™

Catalog No. F056625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F056625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F056625G Supplier TCI America Supplier No. F056625G
Only null left

Chemical Identifiers

CAS 371-15-3
Molecular Formula C7H7FS
Molecular Weight (g/mol) 142.191
MDL Number MFCD00040829
InChI Key XFUMHENRNCUHOH-UHFFFAOYSA-N
Synonym 4-fluorothioanisole, 1-fluoro-4-methylthio benzene, 4-fluorophenyl methyl sulfide, 4-fluoro thioanisole, 1-fluoro-4-methylsulfanyl-benzene, 1-fluoro-4-methylsulfanyl benzene, p-fluoro methylthio benzene, benzene, 1-fluoro-4-methylthio, 4-fluoro-1-methylthiobenzene, pubchem10641
PubChem CID 3083565
IUPAC Name 1-fluoro-4-methylsulfanylbenzene
SMILES CSC1=CC=C(C=C1)F

Specifications

Boiling Point 185°C
Quantity 25 g
Formula Weight 142.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Fluorothioanisole