missing translation for 'onlineSavingsMsg'
Learn More

4-Fluororesorcinol 98.0+%, TCI America™

Catalog No. F07411G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F07411G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F07411G Supplier TCI America Supplier No. F07411G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 103068-41-3
Molecular Formula C6H5FO2
Molecular Weight (g/mol) 128.102
MDL Number MFCD03789074
InChI Key XPOIJNIQXJYQOV-UHFFFAOYSA-N
Synonym 4-fluororesorcinol, 4-fluoro-1,3-benzenediol, 1-fluoro-2,4-dihydroxybenzene, 4-fluoro-1,3-dihydroxybenzene, 1,3-benzenediol, 4-fluoro, 2,4-dihydroxyfluorobenzene, 4fluororesorcinol, pubchem22656, 4-fluoroben-zene-1,3-diol
PubChem CID 10558640
IUPAC Name 4-fluorobenzene-1,3-diol
SMILES C1=CC(=C(C=C1O)O)F

Specifications

Melting Point 97°C
Quantity 1 g
Formula Weight 128.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluororesorcinol