missing translation for 'onlineSavingsMsg'
Learn More

4-Fluororesorcinol, 97%

Catalog No. AAH60426ME
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH60426ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH60426ME Supplier Thermo Scientific Chemicals Supplier No. H60426ME
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 103068-41-3 | C6H5FO2 | 128.102 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 103068-41-3
Molecular Formula C6H5FO2
Molecular Weight (g/mol) 128.102
MDL Number MFCD03789074
InChI Key XPOIJNIQXJYQOV-UHFFFAOYSA-N
Synonym 4-fluororesorcinol, 4-fluoro-1,3-benzenediol, 1-fluoro-2,4-dihydroxybenzene, 4-fluoro-1,3-dihydroxybenzene, 1,3-benzenediol, 4-fluoro, 2,4-dihydroxyfluorobenzene, 4fluororesorcinol, pubchem22656, 4-fluoroben-zene-1,3-diol
PubChem CID 10558640
IUPAC Name 4-fluorobenzene-1,3-diol
SMILES C1=CC(=C(C=C1O)O)F

Specifications

Melting Point 96°C to 100°C
Quantity 500 mg
Formula Weight 128.1
Percent Purity 97%
Chemical Name or Material 4-Fluororesorcinol
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only